Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBSCR17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022479.2 | NP_071924.1 | 11 | 2537 | ||
GenBank mRNA | AF410457.1 | - | 11 | 2271 | ||
AK022143.1 | - | 3 | 805 | |||
AK091379.1 | - | 7 | 1487 | |||
AK094958.1 | - | 11 | 2205 | |||
AK126044.1 | - | 11 | 2673 | |||
AL137431.1 | - | 3 | 878 | |||
CR749675.1 | - | 2 | 638 |