Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022497.4 | NP_071942.1 | 4 | 801 | ||
NR_135246.1 | 4 | 890 | ||||
NR_135247.1 | 4 | 801 | ||||
GenBank mRNA | AK024433.1 | - | 4 | 716 | ||
AK024702.1 | - | 4 | 716 | |||
AK098361.1 | - | 1 | 1612 | |||
BC003590.2 | - | 4 | 751 | 2820823 |