Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193480.1 | NP_001180409.1 | 6 | 1335 | ||
NM_022568.3 | NP_072090.1 | 7 | 1485 | |||
NM_170771.2 | NP_739577.1 | 6 | 1323 | |||
GenBank mRNA | AF303134.1 | - | 7 | 1476 | ||
AK074266.1 | - | 6 | 1316 | |||
AK222848.1 | - | 6 | 1316 | |||
AK290784.1 | - | 6 | 1270 | |||
AK298325.1 | - | 6 | 1297 | |||
BC113862.1 | - | 7 | 1455 | 40033493 | ||
BC114473.1 | - | 7 | 1331 | 40033488 | ||
BX647324.1 | - | 1 | 1066 |