Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282949.2 | NP_001269878.1 | 6 | 1176 | ||
NM_001282950.2 | NP_001269879.1 | 8 | 1350 | |||
NM_001282951.2 | NP_001269880.1 | 5 | 1189 | |||
NM_001282952.2 | NP_001269881.1 | 6 | 1221 | |||
NM_001308381.1 | NP_001295310.1 | 8 | 1365 | |||
NM_022730.3 | NP_073567.1 | 7 | 1297 | |||
XM_006712693.1 | XP_006712756.1 | 8 | 1422 | |||
XM_011511638.1 | XP_011509940.1 | 7 | 2805 | |||
XM_011511639.1 | XP_011509941.1 | 7 | 1238 | |||
XM_011511640.1 | XP_011509942.1 | 7 | 1323 | |||
XM_017004716.1 | XP_016860205.1 | 7 | 1354 | |||
XM_017004717.1 | XP_016860206.1 | 6 | 1255 | |||
GenBank mRNA | AK022674.1 | - | 7 | 1228 | ||
AK024273.1 | - | 6 | 1170 | |||
AK098334.1 | - | 4 | 1634 | |||
AK124133.1 | - | 7 | 1296 | |||
AK126326.1 | - | 7 | 2552 | |||
AK222879.1 | - | 7 | 1235 | |||
BC010739.1 | - | 4 | 832 | |||
BC091493.1 | - | 8 | 1285 | 30387712 | ||
HY051259.1 | - | 2 | 351 |