Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM160B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022749.5 | NP_073586.5 | 13 | 1731 | ||
XM_005273602.3 | XP_005273659.1 | 13 | 1731 | |||
XM_006716381.2 | XP_006716444.1 | 13 | 1731 | |||
XM_006716385.3 | XP_006716448.1 | 14 | 3226 | |||
XM_017013732.1 | XP_016869221.1 | 13 | 1728 | |||
XM_017013733.1 | XP_016869222.1 | 14 | 1848 | |||
XM_017013734.1 | XP_016869223.1 | 13 | 1712 | |||
GenBank mRNA | AF495722.1 | - | 8 | 1138 | ||
AK025454.1 | - | 2 | 141 | |||
AK055191.1 | - | 7 | 790 | |||
AK095924.1 | - | 1 | 21 | |||
BC012865.1 | - | 4 | 544 | |||
BC013350.2 | - | 3 | 222 | |||
BC052237.1 | - | 13 | 1728 | |||
BC112147.1 | - | 7 | 820 |