Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022766.5 | NP_073603.2 | 13 | 1669 | ||
XM_017028909.1 | XP_016884398.1 | 13 | 1466 | |||
GenBank mRNA | AB051433.1 | - | 12 | 1395 | ||
AB079066.1 | - | 13 | 1685 | |||
AJ457828.1 | - | 13 | 1632 | |||
AK291476.1 | - | 13 | 1625 | |||
AL832794.1 | - | 3 | 246 | |||
BC004278.1 | - | 3 | 246 | |||
BC067255.1 | - | 12 | 1546 | |||
BC108712.1 | - | 2 | 171 | |||
BC126940.1 | - | 13 | 1612 | 40084110 | ||
BQ710170.1 | - | 2 | 553 | |||
BU187541.1 | - | 2 | 170 | |||
CR456404.1 | - | 13 | 1572 |