Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201545.1 | NP_001188474.1 | 1 | 483 | ||
NM_022768.4 | NP_073605.4 | 1 | 483 | |||
GenBank mRNA | AF368062.1 | - | 1 | 483 | ||
AF368063.1 | - | 1 | 483 | |||
AF368064.1 | - | 1 | 483 | |||
AK022541.1 | - | 1 | 363 | |||
BC006397.2 | - | 1 | 340 | |||
BC047479.1 | - | 1 | 372 | |||
BC062316.1 | - | 1 | 340 | |||
BC098140.1 | - | 1 | 470 | 40008573 | ||
BC103493.1 | - | 1 | 470 | 40008572 | ||
BC103507.1 | - | 1 | 470 | 40008575 | ||
BC103660.1 | - | 1 | 470 |