Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS8L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022772.3 | NP_073609.2 | 21 | 3112 | ||
XM_017018130.1 | XP_016873619.1 | 22 | 3329 | |||
XM_017018131.1 | XP_016873620.1 | 21 | 3200 | |||
XM_017018132.1 | XP_016873621.1 | 22 | 3170 | |||
GenBank mRNA | AK025588.1 | - | 21 | 3030 | ||
AK025824.1 | - | 21 | 3030 | |||
AK027765.1 | - | 9 | 1924 | |||
AK094539.1 | - | 13 | 2208 | |||
AK122903.1 | - | 18 | 3308 | |||
AK122984.1 | - | 6 | 1578 |