Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297778.1 | NP_001284707.1 | 5 | 871 | ||
NM_022787.3 | NP_073624.2 | 5 | 856 | |||
XM_017002107.1 | XP_016857596.1 | 5 | 1009 | |||
GenBank mRNA | AF312734.1 | - | 4 | 800 | ||
AF314163.1 | - | 4 | 712 | |||
AF459819.1 | - | 4 | 768 | |||
AK315640.1 | - | 5 | 810 | |||
BC014943.1 | - | 5 | 855 | 4874147 |