Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282805.1 | NP_001269734.1 | 5 | 553 | ||
NM_001282806.1 | NP_001269735.1 | 3 | 425 | |||
NM_001282807.1 | NP_001269736.1 | 2 | 300 | |||
NM_022826.3 | NP_073737.1 | 3 | 425 | |||
XM_005246773.2 | XP_005246830.1 | 3 | 448 | |||
XM_005246774.1 | XP_005246831.1 | 3 | 448 | |||
XM_017004718.1 | XP_016860207.1 | 3 | 425 | |||
XM_017004719.1 | XP_016860208.1 | 3 | 451 | |||
XM_017004720.1 | XP_016860209.1 | 3 | 413 | |||
XM_017004721.1 | XP_016860210.1 | 3 | 443 | |||
GenBank mRNA | AK022973.1 | - | 3 | 383 | ||
AK222880.1 | - | 3 | 375 | |||
AK292836.1 | - | 5 | 521 | |||
AK299051.1 | - | 3 | 393 | |||
AK302347.1 | - | 2 | 249 | |||
AK310238.1 | - | 2 | 257 | |||
AK316382.1 | - | 2 | 257 | |||
BC065014.1 | - | 3 | 349 | 6169288 | ||
BI462287.1 | - | 305 | ||||
DA825046.1 | - | 3 | 429 |