Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCLRE1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319946.1 | NP_001306875.1 | 1, 2 | 491 | ||
NM_001319947.1 | NP_001306876.1 | 2, 3 | 405 | |||
NM_022836.3 | NP_073747.1 | 2, 3 | 657 | |||
GenBank mRNA | AF306691.1 | - | 1, 2 | 375 | ||
AF306692.1 | - | 1, 2 | 32 | |||
AF306693.1 | - | 2, 3 | 592 | |||
AK022872.1 | - | 2, 3 | 203 | |||
AK024060.1 | - | 2, 3 | 575 | |||
BC029687.1 | - | 2, 3 | 534 | 5001181 | ||
CR936679.1 | - | 1, 2 | 416 | |||
DA414122.1 | - | 2, 3 | 405 |