Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022917.4 | NP_075068.2 | 3 | 415 | ||
NM_139235.3 | NP_631981.2 | 3 | 415 | |||
XM_017015044.1 | XP_016870533.1 | 3 | 351 | |||
GenBank mRNA | AF361079.1 | - | 3 | 373 | ||
AF361080.1 | - | 3 | 373 | |||
AF361081.1 | - | 3 | 373 | |||
AK026258.1 | - | 3 | 374 | |||
AK293969.1 | - | 3 | 374 | |||
BC020450.1 | - | 3 | 360 | |||
BC030139.2 | - | 3 | 328 | 4423152 |