Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023011.3 | NP_075387.1 | 10 | 1636 | ||
NM_080687.2 | NP_542418.1 | 9 | 1537 | |||
XM_005266202.4 | XP_005266259.1 | 7 | 1262 | |||
XM_006719991.3 | XP_006720054.1 | 9 | 1511 | |||
XM_011534844.1 | XP_011533146.1 | 8 | 1404 | |||
XM_011534845.2 | XP_011533147.1 | 10 | 2164 | |||
XM_011534847.2 | XP_011533149.1 | 9 | 1976 | |||
XM_011534848.2 | XP_011533150.1 | 10 | 2121 | |||
XM_017020709.1 | XP_016876198.1 | 9 | 2348 | |||
XM_017020710.1 | XP_016876199.1 | 10 | 1733 | |||
XM_017020711.1 | XP_016876200.1 | 9 | 1634 | |||
GenBank mRNA | AF318575.1 | - | 10 | 1529 | ||
AK001069.1 | - | 5 | 913 | |||
AK097027.1 | - | 6 | 1598 | |||
AK302003.1 | - | 11 | 1371 | |||
AY013250.1 | - | 10 | 1636 | |||
BC008694.1 | - | 6 | 1049 | |||
BC023569.1 | - | 9 | 1501 | 4127465 |