Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293285.1 | NP_001280214.1 | 10 | 1841 | ||
NM_001293286.1 | NP_001280215.1 | 8 | 1694 | |||
NM_023071.3 | NP_075559.2 | 9 | 1757 | |||
NR_120663.1 | 9 | 1757 | ||||
XM_011538690.2 | XP_011536992.1 | 10 | 1229 | |||
XM_011538691.2 | XP_011536993.1 | 8 | 1124 | |||
XM_017019840.1 | XP_016875329.1 | 9 | 1197 | |||
GenBank mRNA | AB075515.1 | - | 7 | 785 | ||
AK023179.1 | - | 5 | 666 | |||
AK023202.1 | - | 9 | 1156 | |||
AK024678.1 | - | 1 | 53 | |||
AK095582.1 | - | 9 | 1757 | |||
AK292334.1 | - | 8 | 1094 | |||
AL833614.1 | - | 10 | 1282 | |||
BC048299.1 | - | 8 | 1124 | 5520818 | ||
BX648916.1 | - | 8 | 1067 |