Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024045.1 | NP_076950.1 | 5 | 831 | ||
XM_005270148.2 | XP_005270205.1 | 6 | 876 | |||
XM_011540144.2 | XP_011538446.1 | 6 | 883 | |||
XM_017016626.1 | XP_016872115.1 | 5 | 823 | |||
XM_017016627.1 | XP_016872116.1 | 7 | 1299 | |||
XM_017016628.1 | XP_016872117.1 | 6 | 1125 | |||
XM_017016629.1 | XP_016872118.1 | 5 | 1013 | |||
XM_017016630.1 | XP_016872119.1 | 7 | 899 | |||
GenBank mRNA | AB051075.1 | - | 5 | 813 | ||
AF334103.1 | - | 5 | 808 | |||
AK223297.1 | - | 5 | 829 | |||
AK293577.1 | - | 5 | 790 | |||
BC000272.1 | - | 5 | 831 | 3357684 |