Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303039.1 | NP_001289968.1 | 4 | 799 | ||
NM_024067.3 | NP_076972.2 | 4 | 856 | |||
XM_005249845.4 | XP_005249902.1 | 4 | 874 | |||
XM_011515518.2 | XP_011513820.1 | 4 | 874 | |||
XM_017012614.1 | XP_016868103.1 | 4 | 1402 | |||
XM_017012615.1 | XP_016868104.1 | 3 | 398 | |||
GenBank mRNA | AK056149.1 | - | 4 | 593 | ||
BC001076.2 | - | 4 | 675 | 2821816 | ||
BC005121.1 | - | 4 | 675 | 2821816 | ||
CA454040.1 | - | 1 | 13 |