Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111322.1 | NP_001104792.1 | 20 | 4209 | ||
NM_024072.3 | NP_076977.3 | 20 | 4206 | |||
GenBank mRNA | AF478457.1 | - | 20 | 4209 | ||
BC005848.2 | - | 20 | 4127 |