Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039375.2 | NP_001034464.1 | 6 | 538 | ||
NM_024120.4 | NP_077025.2 | 7 | 622 | |||
NR_029377.1 | 7 | 622 | ||||
XM_006723620.3 | XP_006723683.1 | 7 | 599 | |||
XM_006723622.2 | XP_006723685.1 | 6 | 503 | |||
XM_006723623.2 | XP_006723686.1 | 6 | 635 | |||
XM_006723624.2 | XP_006723687.1 | 6 | 649 | |||
XM_011529342.2 | XP_011527644.1 | 7 | 599 | |||
XM_011529344.2 | XP_011527646.1 | 4 | 977 | |||
XM_017028061.1 | XP_016883550.1 | 7 | 734 | |||
GenBank mRNA | AK025977.1 | - | 1 | 48 | ||
AK091060.1 | - | 7 | 599 | |||
AK289781.1 | - | 7 | 614 | |||
BC005984.1 | - | 7 | 432 | |||
BC073158.1 | - | 6 | 515 | 6053315 | ||
CB156360.1 | - | 6 | 508 |