Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161527.1 | NP_001154999.1 | 2 | 337 | ||
NM_001292016.1 | NP_001278945.1 | 2 | 623 | |||
NM_024309.3 | NP_077285.3 | 2 | 623 | |||
GenBank mRNA | AF372839.1 | - | 2 | 623 | ||
AJ304866.1 | - | 2 | 570 | |||
AK026176.1 | - | 2 | 336 | |||
AK096296.1 | - | 2 | 614 | |||
BC002740.2 | - | 2 | 581 | 3632736 | ||
BX386180.2 | - | 2 | 584 | |||
DA421921.1 | - | 2 | 338 |