Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPD52L2 SIRNA Assays ›
Gene Symbol:
TPD52L2
Gene Name:
tumor protein D52 like 2
Gene Aliases:
D54, TPD54
Chromosome Location:
Chr.20: 63865228 - 63891545 on Build GRCh38
UniGene ID:
Hs.473296
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
13057
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TPD52L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243891.1
NP_001230820.1
3
245
NM_001243892.1
NP_001230821.1
3
251
NM_001243894.1
NP_001230823.1
3
320
NM_001243895.1
NP_001230824.1
3
320
NM_003288.3
NP_003279.2
3
320
NM_199359.2
NP_955391.1
3
320
NM_199360.2
NP_955392.1
3
320
NM_199361.2
NP_955393.1
3
320
NM_199362.2
NP_955394.1
3
320
NM_199363.2
NP_955395.1
3
320
NR_045090.1
3
320
XM_011529034.2
XP_011527336.1
3
320
XM_011529035.2
XP_011527337.1
3
320
XM_017028038.1
XP_016883527.1
3
305
GenBank mRNA
AF004430.1
-
3
258
AK055068.1
-
3
279
AK223031.1
-
3
232
AK293349.1
-
3
142
AK297064.1
-
3
278
AK298366.1
-
3
236
AK314501.1
-
3
278
AU124850.1
-
3
249
AY323819.1
-
3
190
BC006804.1
-
3
205
3446037
BI667351.1
-
2
72
BI754201.1
-
3
206
BM473873.1
-
3
277
BM833522.1
-
3
311
BP347669.1
-
3
218
BQ948007.1
-
3
246
BT019631.1
-
3
182
CR542036.1
-
3
182
CR749208.1
-
3
219
DA547106.1
-
3
221
DA689198.1
-
3
219
DA871584.1
-
3
320
DC362382.1
-
3
236
DC428780.1
-
3
175
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_198744, XM_006235761, XM_008762500, XM_006235760, XM_006235757, and others.
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-v99nb:80/100.66.78.103:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline