Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTPAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039199.2 | NP_001034288.1 | 3 | 590 | ||
NM_001261839.1 | NP_001248768.1 | 3 | 590 | |||
NM_024331.4 | NP_077307.2 | 4 | 614 | |||
XM_011529043.2 | XP_011527345.1 | 4 | 726 | |||
XM_011529044.2 | XP_011527346.1 | 4 | 1341 | |||
XM_011529045.2 | XP_011527347.1 | 3 | 590 | |||
XM_017028063.1 | XP_016883552.1 | 4 | 605 | |||
GenBank mRNA | AK314045.1 | - | 3 | 549 | ||
BC003071.2 | - | 4 | 593 | 2989837 | ||
BP351602.1 | - | 3 | 592 | |||
BQ646722.1 | - | 3 | 556 | |||
CR936621.1 | - | 3 | 544 |