Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024525.4 | NP_078801.3 | 5 | 569 | ||
XM_005273260.1 | XP_005273317.1 | 5 | 569 | |||
XM_005273261.2 | XP_005273318.1 | 5 | 569 | |||
XM_005273264.2 | XP_005273321.1 | 5 | 569 | |||
XM_011544275.2 | XP_011542577.1 | 5 | 569 | |||
XM_017002321.1 | XP_016857810.1 | 5 | 569 | |||
XM_017002322.1 | XP_016857811.1 | 5 | 569 | |||
XM_017002325.1 | XP_016857814.1 | 5 | 546 | |||
GenBank mRNA | AK075386.1 | - | 5 | 535 | ||
AL832286.1 | - | 5 | 304 |