Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024546.3 | NP_078822.3 | 4 | 403 | ||
XM_011535225.1 | XP_011533527.1 | 3 | 253 | |||
XM_017020743.1 | XP_016876232.1 | 4 | 2198 | |||
GenBank mRNA | AK098640.1 | - | 4 | 415 | ||
AL136651.1 | - | 4 | 375 | |||
BC136764.1 | - | 4 | 398 | 9020754 | ||
BC136765.1 | - | 4 | 398 | 9020755 | ||
BI550796.1 | - | 4 | 321 | |||
CR936752.1 | - | 4 | 380 | |||
CR936754.1 | - | 4 | 390 | |||
CR936764.1 | - | 4 | 377 |