Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024546.3 | NP_078822.3 | 6 | 3391 | ||
XM_011535225.1 | XP_011533527.1 | 5 | 3241 | |||
XM_017020743.1 | XP_016876232.1 | 6 | 5186 | |||
GenBank mRNA | AK023511.1 | - | 1 | 2578 | ||
AL136651.1 | - | 6 | 3364 | |||
CR936752.1 | - | 6 | 3369 | |||
CR936754.1 | - | 6 | 3379 | |||
CR936764.1 | - | 7 | 3295 |