Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024552.2 | NP_078828.2 | 11 | 1188 | ||
XM_011528290.2 | XP_011526592.1 | 11 | 1224 | |||
XM_011528291.2 | XP_011526593.1 | 11 | 1341 | |||
XM_011528292.2 | XP_011526594.1 | 12 | 2164 | |||
XM_011528293.2 | XP_011526595.1 | 12 | 1340 | |||
XM_011528294.2 | XP_011526596.1 | 12 | 1308 | |||
XM_011528295.2 | XP_011526597.1 | 10 | 994 | |||
XM_017027303.1 | XP_016882792.1 | 12 | 2196 | |||
XM_017027304.1 | XP_016882793.1 | 13 | 1419 | |||
XM_017027305.1 | XP_016882794.1 | 10 | 1055 | |||
GenBank mRNA | AK022151.1 | - | 11 | 1148 | ||
AK055414.1 | - | 8 | 1725 | |||
AK222621.1 | - | 11 | 1155 | |||
BC009828.2 | - | 10 | 991 | 3936998 |