Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258311.1 | NP_001245240.1 | 7 | 2564 | ||
XM_017002330.1 | XP_016857819.1 | 7 | 3633 | |||
GenBank mRNA | AK021475.1 | - | 3 | 1319 | ||
AL833243.1 | - | 6 | 2209 | |||
BC013901.2 | - | 3 | 1373 | |||
BC027466.1 | - | 4 | 1533 | |||
BC036865.1 | - | 6 | 1991 | |||
KF670820.1 | - | 7 | 2365 |