Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCPKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040662.1 | NP_001035752.1 | 5 | 622 | ||
NM_024558.2 | NP_078834.2 | 6 | 727 | |||
NR_049738.1 | 6 | 716 | ||||
NR_049739.1 | 5 | 823 | ||||
GenBank mRNA | AK023982.1 | - | 6 | 716 | ||
AK129564.1 | - | 5 | 751 | |||
BC105118.1 | - | 6 | 737 | 8144121 | ||
BC143673.1 | - | 6 | 748 | 9052187 | ||
BC143674.1 | - | 5 | 643 | 9052188 | ||
BI753530.1 | - | 6 | 707 | |||
BI767189.1 | - | 5 | 593 | |||
BX247997.1 | - | 5 | 802 |