Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324266.1 | NP_001311195.1 | 7 | 1446 | ||
NM_024576.4 | NP_078852.3 | 7 | 1443 | |||
XM_011536117.2 | XP_011534419.1 | 7 | 1379 | |||
XM_011536118.2 | XP_011534420.1 | 7 | 1228 | |||
XM_017011303.1 | XP_016866792.1 | 7 | 1008 | |||
XM_017011304.1 | XP_016866793.1 | 7 | 1005 | |||
XM_017011305.1 | XP_016866794.1 | 7 | 901 | |||
XM_017011306.1 | XP_016866795.1 | 7 | 1931 | |||
GenBank mRNA | AK024732.1 | - | 4 | 506 | ||
AK308623.1 | - | 3 | 524 | |||
BC018688.1 | - | 7 | 1078 | 4717736 | ||
BC110999.1 | - | 7 | 1082 | 5585901 |