Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024594.3 | NP_078870.1 | 2 | 604 | ||
GenBank mRNA | AK022961.1 | - | 2 | 349 | ||
AK223285.1 | - | 2 | 333 | |||
BC013705.1 | - | 2 | 578 | 4343270 |