Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QTRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256835.1 | NP_001243764.1 | 9 | 2145 | ||
NM_001256836.1 | NP_001243765.1 | 6 | 1791 | |||
NM_001256837.1 | NP_001243766.1 | 8 | 1889 | |||
NM_024638.3 | NP_078914.1 | 10 | 2253 | |||
XM_005247769.3 | XP_005247826.1 | 10 | 2034 | |||
XM_005247770.3 | XP_005247827.1 | 8 | 2017 | |||
XM_006713744.3 | XP_006713807.1 | 9 | 1886 | |||
XM_011513137.2 | XP_011511439.1 | 10 | 2157 | |||
XM_017007182.1 | XP_016862671.1 | 9 | 2107 | |||
XM_017007183.1 | XP_016862672.1 | 7 | 1872 | |||
XM_017007184.1 | XP_016862673.1 | 10 | 2148 | |||
GenBank mRNA | AK023022.1 | - | 10 | 2143 | ||
AL833709.1 | - | 9 | 2113 | |||
BC034559.2 | - | 9 | 2127 | 4385906 | ||
BC039265.1 | - | 10 | 2224 | 5296335 |