Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024641.3 | NP_078917.2 | 2 | 575 | ||
XM_005267147.3 | XP_005267204.1 | 2 | 968 | |||
GenBank mRNA | AJ577574.1 | - | 1 | 440 | ||
AK022900.1 | - | 1 | 81 | |||
AK291940.1 | - | 2 | 506 | |||
AY048774.1 | - | 2 | 530 | |||
AY048775.1 | - | 2 | 530 | |||
AY372528.1 | - | 1 | 395 | |||
BC137014.1 | - | 1 | 444 | 9021007 | ||
BC137016.1 | - | 1 | 444 | 9021009 | ||
BC146671.1 | - | 2 | 510 | 40148201 | ||
BC150199.1 | - | 2 | 510 | 40148811 | ||
BX481372.1 | - | 2 | 579 | |||
BX640869.1 | - | 2 | 536 | |||
CR936827.1 | - | 2 | 510 |