Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024715.3 | NP_078991.3 | 2, 3 | 1257 | ||
GenBank mRNA | AK026278.1 | - | 3, 4 | 660 | ||
AK056512.1 | - | 2, 3 | 1292 | |||
AK092425.1 | - | 1 | 1322 | |||
AK225454.1 | - | 3, 4 | 660 | |||
AK225462.1 | - | 3, 4 | 660 | |||
AK299578.1 | - | 2, 3 | 550 | |||
AY358649.1 | - | 2, 3 | 685 | |||
BC001615.1 | - | 2, 3 | 744 | 3544923 | ||
BC032568.1 | - | 2, 3 | 658 | 5225890 | ||
BX538131.1 | - | 2, 3 | 584 |