Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024715.3 | NP_078991.3 | 4 | 1474 | ||
GenBank mRNA | AK026278.1 | - | 5 | 877 | ||
AK056512.1 | - | 4 | 1509 | |||
AK092425.1 | - | 2 | 1539 | |||
AK225454.1 | - | 5 | 877 | |||
AK225462.1 | - | 5 | 877 | |||
AK299578.1 | - | 4 | 767 | |||
AK307529.1 | - | 3 | 385 | |||
AY358649.1 | - | 4 | 902 | |||
BC001615.1 | - | 4 | 961 | 3544923 | ||
BC032568.1 | - | 4 | 875 | 5225890 | ||
BX538131.1 | - | 4 | 801 |