Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286732.1 | NP_001273661.1 | 7 | 873 | ||
NM_024719.3 | NP_078995.2 | 7 | 873 | |||
XM_005268333.3 | XP_005268390.1 | 6 | 710 | |||
XM_017020755.1 | XP_016876244.1 | 7 | 831 | |||
XM_017020756.1 | XP_016876245.1 | 8 | 1011 | |||
GenBank mRNA | AB449887.1 | - | 7 | 776 | ||
AK026127.1 | - | 5 | 516 | |||
AK300897.1 | - | 7 | 823 | |||
BC033071.2 | - | 5 | 597 | |||
BC062461.1 | - | 6 | 721 | |||
BC080640.1 | - | 7 | 830 | |||
BC112123.1 | - | 5 | 550 | 8327591 | ||
BC112125.1 | - | 5 | 562 | 8327593 | ||
BC143682.1 | - | 4 | 622 | |||
CX788865.1 | - | 5 | 786 |