Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286732.1 | NP_001273661.1 | 7 | 908 | ||
NM_024719.3 | NP_078995.2 | 7 | 908 | |||
XM_005268333.3 | XP_005268390.1 | 6 | 745 | |||
XM_017020755.1 | XP_016876244.1 | 7 | 866 | |||
XM_017020756.1 | XP_016876245.1 | 8 | 1046 | |||
GenBank mRNA | AB449887.1 | - | 7 | 811 | ||
AK026127.1 | - | 5 | 551 | |||
AK300897.1 | - | 7 | 858 | |||
BC033071.2 | - | 5 | 632 | |||
BC062461.1 | - | 6 | 756 | |||
BC080640.1 | - | 7 | 865 | |||
BC112123.1 | - | 5 | 585 | 8327591 | ||
BC112125.1 | - | 5 | 597 | 8327593 | ||
BC143682.1 | - | 4 | 657 | |||
CX788865.1 | - | 5 | 821 |