Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144766.2 | NP_001138238.1 | 3 | 326 | ||
NM_003315.3 | NP_003306.3 | 3 | 558 | |||
NR_029431.1 | 2 | 469 | ||||
XM_011525167.2 | XP_011523469.1 | 4 | 488 | |||
XM_011525168.2 | XP_011523470.1 | 4 | 448 | |||
XM_011525169.2 | XP_011523471.1 | 3 | 300 | |||
XM_017024994.1 | XP_016880483.1 | 4 | 482 | |||
GenBank mRNA | AB209834.1 | - | 3 | 244 | ||
AK291047.1 | - | 3 | 259 | |||
AK296602.1 | - | 3 | 280 | |||
AK298860.1 | - | 3 | 274 | |||
AK308016.1 | - | 3 | 312 | |||
AK316081.1 | - | 2 | 195 | |||
BC003601.1 | - | 2 | 181 | 3543074 | ||
BC011837.2 | - | 3 | 277 | 3945721 | ||
BC033772.1 | - | 3 | 303 | 3924081 | ||
BP378441.1 | - | 3 | 307 | |||
BX647209.1 | - | 3 | 514 | |||
U46571.1 | - | 3 | 273 |