Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001894.1 | NP_001001894.1 | 6 | 561 | ||
NM_001320703.1 | NP_001307632.1 | 6 | 767 | |||
NM_001320704.1 | NP_001307633.1 | 6 | 757 | |||
NM_003316.3 | NP_003307.3 | 6 | 1852 | |||
XM_005261048.1 | XP_005261105.1 | 6 | 619 | |||
XM_005261049.1 | XP_005261106.1 | 6 | 542 | |||
XM_005261050.4 | XP_005261107.1 | 6 | 3504 | |||
XM_005261052.1 | XP_005261109.1 | 6 | 542 | |||
XM_005261054.3 | XP_005261111.1 | 6 | 3496 | |||
XM_011529740.2 | XP_011528042.1 | 5 | 467 | |||
XM_011529742.2 | XP_011528044.2 | 6 | 564 | |||
XM_017028458.1 | XP_016883947.1 | 6 | 558 | |||
XM_017028459.1 | XP_016883948.1 | 6 | 557 | |||
XM_017028460.1 | XP_016883949.1 | 6 | 556 | |||
XM_017028461.1 | XP_016883950.1 | 6 | 619 | |||
XM_017028462.1 | XP_016883951.1 | 6 | 542 | |||
XM_017028463.1 | XP_016883952.1 | 6 | 619 | |||
XM_017028465.1 | XP_016883954.1 | 2 | 118 | |||
GenBank mRNA | AJ001866.1 | - | 3 | 613 | ||
AK125473.1 | - | 6 | 3204 | |||
AK291992.1 | - | 6 | 669 | |||
BC063033.1 | - | 6 | 579 | |||
BC092466.1 | - | 6 | 564 | |||
BC137345.1 | - | 6 | 991 | 9021343 | ||
D83077.1 | - | 6 | 1852 | |||
D83327.1 | - | 3 | 204 | |||
D84294.1 | - | 5 | 1925 | |||
D84295.1 | - | 5 | 1925 |