Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPERB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024764.3 | NP_079040.2 | 19 | 2252 | ||
GenBank mRNA | AK024360.1 | - | 5 | 780 | ||
AK126034.1 | - | 19 | 2236 | |||
BC126214.1 | - | 19 | 2206 | 8991930 | ||
BC126216.1 | - | 19 | 2206 | 8991932 |