Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP4K2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146258.1 | NP_001139730.1 | 10 | 1357 | ||
NM_001146259.1 | NP_001139731.1 | 10 | 1303 | |||
NM_001146260.1 | NP_001139732.1 | 9 | 1213 | |||
NM_024779.4 | NP_079055.3 | 10 | 1357 | |||
XM_005269152.2 | XP_005269209.1 | 9 | 1311 | |||
XM_017019973.1 | XP_016875462.1 | 11 | 1419 | |||
GenBank mRNA | AK025708.1 | - | 6 | 636 | ||
AK075553.1 | - | 10 | 1312 | |||
AK125526.1 | - | 10 | 1241 | |||
AK225343.1 | - | 6 | 636 | |||
AK225685.1 | - | 6 | 644 | |||
AK297243.1 | - | 9 | 1198 | |||
AK302254.1 | - | 10 | 1219 | |||
AK315588.1 | - | 10 | 1237 | |||
AX754970.1 | - | 10 | 1298 | |||
BC028596.2 | - | 10 | 1242 | 4824916 |