Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024784.3 | NP_079060.1 | 2 | 1811 | ||
XM_011545256.2 | XP_011543558.1 | 2 | 1633 | |||
GenBank mRNA | AK027045.1 | - | 2 | 1786 | ||
BC025249.1 | - | 2 | 1769 | 5444179 |