Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024786.2 | NP_079062.1 | 5 | 1081 | ||
XM_017009869.1 | XP_016865358.1 | 6 | 962 | |||
XM_017009870.1 | XP_016865359.1 | 8 | 2773 | |||
XM_017009871.1 | XP_016865360.1 | 8 | 2722 | |||
XM_017009872.1 | XP_016865361.1 | 6 | 797 | |||
XM_017009873.1 | XP_016865362.1 | 2 | 779 | |||
XM_017009874.1 | XP_016865363.1 | 2 | 1659 | |||
XM_017009875.1 | XP_016865364.1 | 2 | 1606 | |||
XM_017009876.1 | XP_016865365.1 | 2 | 573 | |||
XM_017009877.1 | XP_016865366.1 | 2 | 147 | |||
XM_017009878.1 | XP_016865367.1 | 2 | 104 | |||
GenBank mRNA | AK023215.1 | - | 5 | 1081 | ||
AK024233.1 | - | 2 | 1762 | |||
AY358673.1 | - | 2 | 163 | |||
BC032000.1 | - | 5 | 700 | 4750082 |