Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024805.2 | NP_079081.2 | 5 | 690 | ||
GenBank mRNA | AF261073.2 | - | 5 | 570 | ||
AK024825.1 | - | 5 | 678 | |||
BC014195.2 | - | 5 | 552 | 3938406 | ||
BC057772.1 | - | 5 | 558 | 30337652 |