Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136041.2 | NP_001129513.1 | 8 | 622 | ||
NM_024827.3 | NP_079103.2 | 8 | 723 | |||
XM_005265491.1 | XP_005265548.1 | 8 | 688 | |||
XM_005265492.1 | XP_005265549.1 | 5 | 530 | |||
XM_011534131.1 | XP_011532433.1 | 8 | 811 | |||
XM_011534132.2 | XP_011532434.2 | 5 | 376 | |||
XM_011534135.1 | XP_011532437.1 | 7 | 530 | |||
GenBank mRNA | AK025426.1 | - | 8 | 614 | ||
AK025890.1 | - | 8 | 614 | |||
AK293223.1 | - | 8 | 555 | |||
AK299021.1 | - | 4 | 294 | |||
AL834223.1 | - | 7 | 938 | |||
BC009676.1 | - | 7 | 599 | 3906049 | ||
BG708402.1 | - | 8 | 695 | |||
CR457341.1 | - | 4 | 224 |