Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142555.2 | NP_001136027.1 | 8 | 679 | ||
NM_024876.3 | NP_079152.3 | 9 | 1047 | |||
GenBank mRNA | BC013114.1 | - | 9 | 857 | 4342891 | |
BC027473.1 | - | 8 | 646 | 4997823 |