Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142555.2 | NP_001136027.1 | 14 | 1812 | ||
NM_024876.3 | NP_079152.3 | 15 | 2180 | |||
GenBank mRNA | AF052122.1 | - | 4 | 774 | ||
AK057199.1 | - | 5 | 1993 | |||
BC013114.1 | - | 15 | 1990 | 4342891 | ||
BC027473.1 | - | 14 | 1779 | 4997823 |