Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122841.2 | NP_001116313.1 | 11 | 2835 | ||
NM_024919.4 | NP_079195.3 | 11 | 2985 | |||
NR_110312.1 | 16 | 3827 | ||||
XM_011536137.1 | XP_011534439.1 | 13 | 3134 | |||
XM_011536138.1 | XP_011534440.1 | 13 | 2950 | |||
XM_011536140.1 | XP_011534442.1 | 12 | 2989 | |||
XM_011536141.1 | XP_011534443.1 | 12 | 2929 | |||
XM_011536142.2 | XP_011534444.1 | 12 | 2967 | |||
XM_011536143.1 | XP_011534445.1 | 12 | 2865 | |||
XM_011536145.1 | XP_011534447.1 | 12 | 2904 | |||
XM_011536146.2 | XP_011534448.1 | 8 | 3757 | |||
GenBank mRNA | AK026268.1 | - | 3 | 1727 | ||
AK074110.1 | - | 12 | 4657 | |||
AK097179.1 | - | 11 | 2988 | |||
AK097579.1 | - | 11 | 2834 | |||
AK125963.1 | - | 16 | 3799 | |||
AL133077.1 | - | 9 | 2607 |