Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256599.1 | NP_001243528.1 | 9 | 2036 | ||
NM_001256600.1 | NP_001243529.1 | 10 | 1980 | |||
NM_001256601.1 | NP_001243530.1 | 9 | 1867 | |||
NM_001282378.1 | NP_001269307.1 | 8 | 1764 | |||
NM_025058.4 | NP_079334.3 | 9 | 2026 | |||
NR_046327.1 | 10 | 2064 | ||||
NR_046329.1 | 11 | 2123 | ||||
XM_011510002.1 | XP_011508304.1 | 10 | 1969 | |||
XM_017002393.1 | XP_016857882.1 | 10 | 2140 | |||
GenBank mRNA | AK096158.1 | - | 9 | 1914 | ||
AK128139.1 | - | 11 | 2123 | |||
AK296300.1 | - | 10 | 1976 | |||
AK297318.1 | - | 8 | 1672 | |||
AK308075.1 | - | 9 | 1831 | |||
AK309699.1 | - | 9 | 1867 | |||
AY251386.1 | - | 9 | 1846 | |||
BC126372.1 | - | 10 | 1981 | 8992088 | ||
BC143828.1 | - | 10 | 1981 |