Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256599.1 | NP_001243528.1 | 10 | 2734 | ||
NM_001256600.1 | NP_001243529.1 | 11 | 2678 | |||
NM_001256601.1 | NP_001243530.1 | 10 | 2565 | |||
NM_001282378.1 | NP_001269307.1 | 9 | 2462 | |||
NM_025058.4 | NP_079334.3 | 10 | 2724 | |||
NR_046327.1 | 11 | 2762 | ||||
NR_046329.1 | 12 | 2821 | ||||
XM_011510002.1 | XP_011508304.1 | 11 | 2667 | |||
XM_017002393.1 | XP_016857882.1 | 11 | 2838 | |||
GenBank mRNA | AK090504.1 | - | 2 | 1331 | ||
AK096158.1 | - | 10 | 2612 | |||
AK128139.1 | - | 12 | 2821 | |||
AK297318.1 | - | 9 | 2370 |