Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300795.1 | NP_001287724.1 | 8 | 999 | ||
NM_003340.6 | NP_003331.1 | 8 | 1306 | |||
NM_181886.3 | NP_871615.1 | 8 | 1514 | |||
NM_181887.2 | NP_871616.1 | 8 | 1284 | |||
NM_181888.3 | NP_871617.1 | 8 | 1163 | |||
NM_181889.2 | NP_871618.1 | 8 | 1186 | |||
NM_181890.2 | NP_871619.1 | 8 | 1128 | |||
NM_181891.2 | NP_871620.1 | 8 | 1471 | |||
NM_181892.3 | NP_871621.1 | 8 | 1546 | |||
NM_181893.2 | NP_871622.1 | 7 | 976 | |||
XM_017008584.1 | XP_016864073.1 | 7 | 1338 | |||
GenBank mRNA | AK095822.1 | - | 8 | 1461 | ||
AK127304.1 | - | 6 | 3919 | |||
BC003395.1 | - | 8 | 986 | 3447673 | ||
BC037894.2 | - | 8 | 1284 | 5275356 | ||
BC066917.1 | - | 8 | 1017 | 4792462 |