Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASRGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083926.1 | NP_001077395.1 | 7 | 1297 | ||
NM_025080.3 | NP_079356.3 | 7 | 1220 | |||
XM_005274305.3 | XP_005274362.1 | 6 | 937 | |||
XM_005274306.2 | XP_005274363.1 | 6 | 999 | |||
GenBank mRNA | AF411076.1 | - | 7 | 1029 | ||
AK090908.1 | - | 7 | 1069 | |||
AK091894.1 | - | 7 | 1151 | |||
AK225802.1 | - | 7 | 988 | |||
BC006267.2 | - | 4 | 610 | |||
BC021295.2 | - | 7 | 1018 | 3937243 | ||
BC064963.1 | - | 6 | 910 | 6141919 | ||
BC093070.1 | - | 7 | 1122 | 30370741 | ||
BX640832.1 | - | 1 | 3317 |